Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KMT3B/NSD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KMT3B/NSD1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KMT3B/NSD1 Polyclonal specifically detects KMT3B/NSD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
KMT3B/NSD1 | |
Polyclonal | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Cellular Markers, Tumor Suppressors | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
64324 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSDVHFDSKVKQSDPGKISEKGLSFENGKGPELDSVMNSENDELNGVNQVVPKKRWQRLNQRRTKPRKRMNRFKEKENSEC | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
Androgen receptor coactivator 267 kDa protein, Androgen receptor-associated protein of 267 kDa, ARA267STO, DKFZp666C163, EC 2.1.1.43, FLJ10684, FLJ22263, FLJ44628, H3 lysine-36 and H4 lysine-20 specific, KMT3BSotos syndrome, Lysine N-methyltransferase 3B, NR-binding SET domain-containing protein, nuclear receptor binding SET domain protein 1 | |
NSD1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title