Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KN-93
GREENER_CHOICESDP

Catalog No. 12781 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-781 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-781 Supplier Tocris Bioscience™ Supplier No. 1278/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaM kinase II inhibitor; also K+ channel blocker (KV)

Potent, cell permeable inhibitor of CaM kinase II (IC50 = 0.37 μM). Also a direct extracellular open channel blocker of voltage-gated potassium channels (IC50 = 307 nM for KV1.5) and abolishes IKr in ventricular myocytes (IC50 = 102.6 nM) independently of CaM kinase II inhibition.

Water soluble form and Negative Control also available.

Chemical Identifiers

CAS 139298-40-1
Molecular Formula C26H29ClN2O4S

Specifications

Quantity 1 mg
Formula Weight 501.04
Purity >98%
Molecular Weight (g/mol) 501.038
Chemical Name or Material KN-93
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.