Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KNAT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144536
Description
KNAT3 Polyclonal Antibody for Western Blot, ELISA
KNAT3 (KNOTTED1-like homeobox protein 3) is a gene that encodes a transcription factor protein. Transcription factors are proteins that regulate the expression of other genes by binding to specific DNA sequences and either promoting or inhibiting their transcription. KNAT3 is a member of the KNOX (KNOTTED1-like homeobox) family of transcription factors, which are involved in various developmental processes, including meristem formation and organogenesis. The specific function of KNAT3 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease.
Specifications
| KNAT3 | |
| Polyclonal | |
| Unconjugated | |
| KNAT3 | |
| Homeobox protein knotted-1-like 3KNAT3 At5g25220 F21J6.18 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 832593 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2.66 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P48000 | |
| KNAT3 | |
| Recombinant Arabidopsis thaliana KNAT3 protein (108-154aa). | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction