Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNAT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144535 Supplier Invitrogen™ Supplier No. PA5144535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KNAT5 Polyclonal Antibody for Western Blot, ELISA

KNAT5 (KNOTTED1-like homeobox protein 5) is a gene that encodes a transcription factor protein. Transcription factors are proteins that regulate the expression of other genes by binding to specific DNA sequences and either promoting or inhibiting their transcription. KNAT5 is a member of the KNOX (KNOTTED1-like homeobox) family of transcription factors, which are involved in various developmental processes, including meristem formation and organogenesis. The specific function of KNAT5 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNAT5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene KNAT5
Gene Accession No. P48002
Gene Alias ATH1
Gene Symbols KNAT5
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana KNAT5 protein (1-118aa).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 829335
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.