Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139829 Supplier Invitrogen™ Supplier No. PA5139829
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ERLVTEKASV YCVAAVLWTA AKFSVPRNHK LALPRRLKTL LLDMARRSAP ERPSAAEAIK VCGSYLLQRG MDSRKILAHL RASICQVYQ Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

The protein encoded by this gene is a Ras guanine nucleotide exchange factor that appears to negatively regulate dendritic growth in the brain. Knockdown of this gene in senescent umbilical vein endothelial cells partially reversed the senescence, showing that this gene could potentially be targeted by anti-aging therapies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNDC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KNDC1
Gene Accession No. Q76NI1
Gene Alias 2410012C07Rik; B830014K08Rik; bB439H18.3; C10orf23; Cerebral protein 9; hucep-9; KIAA1768; kinase non-catalytic C-lobe domain (KIND) containing 1; kinase non-catalytic C-lobe domain containing 1; kinase non-catalytic C-lobe domain-containing protein 1; kind; KIND domain-containing protein 1; Kndc1; Protein very KIND; RasGEF domain family, member 2; ras-GEF domain-containing family member 2; RASGEF2; RGD1565261; very-kind; VKIND; v-KIND
Gene Symbols KNDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KNDC1. Recombinant protein control fragment (Product #RP-108927).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85442
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.