Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KNDC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980652
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0652 Supplier Bioss Supplier No. BS10056R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a Ras guanine nucleotide exchange factor that appears to negatively regulate dendritic growth in the brain. Knockdown of this gene in senescent umbilical vein endothelial cells partially reversed the senescence, showing that this gene could potentially be targeted by anti-aging therapies.

Specifications

Antigen KNDC1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KNDC1
Gene Accession No. Q76NI1
Gene Alias bB439H18.3; C10orf23; Cerebral protein 9; hucep-9; KIAA1768; kinase non-catalytic C-lobe domain (KIND) containing 1; kinase non-catalytic C-lobe domain containing 1; Kinase non-catalytic C-lobe domain-containing protein 1; KIND domain-containing protein 1; KNDC1; Protein very KIND; RasGEF domain family, member 2; Ras-GEF domain-containing family member 2; RASGEF2; VKIND; v-KIND
Gene Symbols KNDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KNDC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85442
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.