Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KNF™ FLEXPAK Polyethylene Antistatic Cleanroom Tubing

Click to view available options
Width (English):
4 in.
6 in.
8 in.
10 in.
18 in.
24 in.
36 in.
48 in.
Length (English):
250 ft.
500 ft.
Description
Constructed from 6mil thick, low-density polyethylene. Treated with an amine-free agent.
- Tough yet flexible
- Chemically resistant to solvents
- Exhibit low water vapor transmission rate and good electrical properties
Order Info
Cleaned and packaged in a Class 10/100 cleanroom, and certified to NASA JPG standards. FDA approved for use with food.
Compliance
Suitable for Class 100 cleanroom environments.
Specifications
Specifications
| For Use With (Application) | Effective for secondary packaging for parts transport |
| Material | Polyethylene |
| Length (Metric) | 76 m |
| Width (English) | 18 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction