Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KNF™ FLEXPAK Polyethylene Ultraclean™ Bags - 6mil Thickness

Click to view available options
Length (English):
3 in.
4 in.
8 in.
10 in.
12 in.
15 in.
Width (English):
3 in.
4 in.
6 in.
10 in.
12 in.
15 in.
24 in.
26 in.
Specifications
Specifications
| Material | LDPE |
| Thickness (English) | 6 mil |
| Length (English) | 10 in. |
| Width (English) | 12 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction