Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Knitted Reactor Coil
For use with PHRED Reactor
Supplier: MilliporeSigma™ Supelco™ 57410U
Description

Specifications
L I.D. 20 m 0.25 mm | |
PHRED Reactor | |
1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction