Learn More
MilliporeSigma™ Supelco™ Knitted Reactor Coil
For use with PHRED Reactor
$278.00 - $962.00
Specifications
For Use With (Equipment) PHRED Reactor
View More Specs
Products 8
Catalog Number Mfr. No. Volume (Metric) Description Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Description Price Quantity & Availability  
11-100-4251
View Documents
MilliporeSigma™ Supelco™
57402
0.25 mL L I.D. 5 m 0.25 mm
Each for $278.00
Only null left
Add to Cart
11-100-3827
View Documents
MilliporeSigma™ Supelco™
57403
0.5 mL L I.D. 10 m 0.25 mm
Each for $534.00
Only null left
Add to Cart
11-100-3672
View Documents
MilliporeSigma™ Supelco™
57404
0.75 mL L I.D. 15 m 0.25 mm
Each for $723.00
Only null left
Add to Cart
11-101-6300
View Documents
MilliporeSigma™ Supelco™
57405
1 mL L I.D. 5 m 0.5 mm
Each for $368.00
Only null left
Add to Cart
11-101-6184
View Documents
MilliporeSigma™ Supelco™
57410U
1 mL L I.D. 20 m 0.25 mm
Each for $962.00
Only null left
Add to Cart
11-100-3749
View Documents
MilliporeSigma™ Supelco™
57406
2 mL L I.D. 10 m 0.5 mm
Each for $670.00
Only null left
Add to Cart
11-100-3496
View Documents
MilliporeSigma™ Supelco™
57407
3 mL L I.D. 15 m 0.5 mm
Each for $883.00
Only null left
Add to Cart
11-100-3500
View Documents
MilliporeSigma™ Supelco™
57411
4 mL L I.D. 20 m 0.5 mm
Each for $869.00
Only null left
Add to Cart
Description
Description
Specifications
Specifications
PHRED Reactor
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.