Learn More
Corning™ PYREX™ Knob Top Nonvacuum Glass Desiccators, Complete (Bowl and Cover)
GSA_VA
Strong, easily cleaned, borosilicate glass desiccators designed for general analytical work. The sturdy knobs make the covers easy to handle.
$481.00 - $1102.00
Specifications
Material Borosilicate Glass 7740
Product Type Nonvacuum Desiccator
Style Knob Top
View More Specs
Alert:
Not for vacuum applications. Do not heat or subject to pressure.
Products 3
Catalog Number Mfr. No. Height (Metric) Depth (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Depth (Metric) Size Price Quantity & Availability  
08-624-414
View Documents
Corning™
3081200
30.9 cm 11.5 cm Large
Case for $885.00
Only null left
Add to Cart
08-624-411
GSA_VA
View Documents
Corning™
3081150
25.2 cm 8.1 cm Small
Case for $481.00
Only null left
Add to Cart
08-624-417
GSA_VA
View Documents
Corning™
3081250
35.7 cm 12 cm Large
Case for $1,102.00
Only null left
Add to Cart
Description
Description
PYREX™ desiccators are designed for general analytical work. The sturdy knob makes the cover easy to handle. For replacement cover, see No. 3081-150CO. For lower part only, see No. 3081-150LO. Warning: Do not heat or subject to pressure. Not for vacuum applications.
Order Info
Replacement bowls and covers available separately. See Corning PYREX Brand Desiccator Replacement Covers and Corning PYREX Brand Desiccator Replacement Bowls.
Specifications
Specifications
Borosilicate Glass 7740
Nonvacuum Desiccator
Knob Top
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.