Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNOP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567600 Supplier Invitrogen™ Supplier No. PA567600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KNOP1 Polyclonal Antibody for IHC (P)

KNOP1 is a nucleolar protein that interacts with zinc finger 106 protein. The encoded protein has several of the same characteristics as nucleostemin and may be involved in testis development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNOP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KNOP1
Gene Accession No. Q1ED39
Gene Alias 101F10.1; C16orf88; FAM191A; family with sequence similarity 191, member A; KNOP1; lysine rich nucleolar protein 1; lysine-rich nucleolar protein 1; protein C16orf88; Protein FAM191A; Testis-specific gene 118 protein; TSG118
Gene Symbols KNOP1
Host Species Rabbit
Immunogen Synthesized peptide derived from human KNOP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400506
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.