Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNP-I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553828 Supplier Invitrogen™ Supplier No. PA553828
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGRTPSQRAA LHLSVPRPAA RVALVLSGCG VYDGTEIHEA SAILVHLSRG GAEVQIFAPD VPQMHVIDHT KGQPSEGESR N Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

This gene encodes a potential mitochondrial protein that is a member of the DJ-1/PfpI gene family. This protein is overexpressed in fetal Down syndrome brain. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNP-I
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GATD3A
Gene Accession No. P0DPI2
Gene Alias C21orf33; chromosome 21 open reading frame 33; ES1; ES1 protein homolog, mitochondrial; GATD3A; GATD3B; Glutamine amidotransferase-like class 1 domain-containing protein 3A, mitochondrial; Glutamine amidotransferase-like class 1 domain-containing protein 3B, mitochondrial; GT335; H KNPH; HES1; human HES1 protein, homolog to E.coli and zebrafish ES1 protein; Keio novel protein I; Keio novel protein-I; KNPH; KNPI; KNP-I; Protein GT335; Protein HES1; testis secretory sperm-binding protein Li 237E
Gene Symbols GATD3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KNP-I. Recombinant protein control fragment (Product #RP-91785).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8209
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.