Learn More
Invitrogen™ KNP-I Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109465
Description
Immunogen sequence: AARVALVLSG CGVYDGTEIH EASAILVHLS RGGAEVQIFA PDVPQMHVID HTKGQPSEGE SRNVLTESAR IARGKITDLA NLSAANHDAA IFPGGFGAAK NLSTFAVDGK DCKVNKEVER VLKEFHQAGK PIGLCCIAPV LAAKVLRGVE VTVGHEQEEG GKWPYAGTAE AIKALGAKHC VKEVVEAHVD QKNKVVTTPA FMCETALHYI HDGIGAMVRK VLELTGK.
This gene encodes a potential mitochondrial protein that is a member of the DJ-1/PfpI gene family. This protein is overexpressed in fetal Down syndrome brain. Alternate splicing results in multiple transcript variants.
Specifications
| KNP-I | |
| Polyclonal | |
| Unconjugated | |
| GATD3A | |
| C21orf33; chromosome 21 open reading frame 33; ES1; ES1 protein homolog, mitochondrial; GATD3A; GATD3B; Glutamine amidotransferase-like class 1 domain-containing protein 3A, mitochondrial; Glutamine amidotransferase-like class 1 domain-containing protein 3B, mitochondrial; GT335; H KNPH; HES1; human HES1 protein, homolog to E.coli and zebrafish ES1 protein; Keio novel protein I; Keio novel protein-I; KNPH; KNPI; KNP-I; Protein GT335; Protein HES1; testis secretory sperm-binding protein Li 237E | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8209 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.92 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0DPI2 | |
| GATD3A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-268 of human C21orf33 (NP_004640.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.