Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNT1 Polyclonal Antibody

Catalog No. PIPA5122191
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122191 20 μL
PIPA5119465 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122191 Supplier Invitrogen™ Supplier No. PA5122191
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KNT1 Polyclonal Antibody for Western Blot, IHC (P)

KNT1 (T-Kinninogen 1) is the acute phase kininogen of the rat. KNT1 could be a healing protein because of its properties as a cysteine protease inhibitor. Evidence has been presented that mRNA of T-kininogen synthesis may be a function of interleukin 6 production. In rats, four types of kininogens are produced, two of which are classical high and low molecular weight kininogens and two of which are low molecular weight-like kininogens, designated T-kininogen 1 and T-kininogen 2. T-kininogen 1 and T-kininogen 2 are 430 amino acid secreted rat proteins that each contain three cystatin domains and have nearly identical functions. Existing in plasma, both T-kininogen 1 and T-kininogen 2 are glycoproteins that act as thiol protease inhibitors and also play a role in blood coagulation, specifically by helping to optimally position blood coagulation factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Kng1
Gene Accession No. P08934
Gene Alias T-Kinninogen 1
Gene Symbols Kng1
Host Species Rabbit
Immunogen Recombinant protein T-Kininogen 1. The antigen corresponds to amino acid range 378-430 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288001
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.