Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KNTC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10088130
Description
KNTC1 Polyclonal specifically detects KNTC1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
KNTC1 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
FLJ36151, hRod, hsROD, KIAA0166Rough Deal homolog, centromere/kinetochore protein, kinetochore associated 1, kinetochore-associated protein 1, Rod, RODHsROD, Rough deal homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
NP_055523.1 | |
KNTC1 | |
The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human rough deal homolog using the numbering given in entry NP_055523.1 (GeneID 9735). | |
0.1 mL | |
Mitotic Regulators | |
9735 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction