Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KNTC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555234
Description
Immunogen sequence: VKMLESLLNS MSASVSLQKL CPWFKNDVIP FVRRTVPEGQ IILAKWLEQA ARNLELTDKA NWPENGLQLA EIFFTAEKTD ELGLASSWHW ISLKDYQNTE EVCQLRTLVN NLR Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
Specifications
| KNTC1 | |
| Polyclonal | |
| Unconjugated | |
| KNTC1 | |
| D330012D13; FLJ36151; HsROD; Kiaa0166; kinetochore associated 1; kinetochore-associated protein 1; Kinetochore-associated protein 1-like protein; KNTC1; LOW QUALITY PROTEIN: kinetochore-associated protein 1; ROD; Rough deal homolog; Rough Deal homolog, centromere/kinetochore protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9735 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50748 | |
| KNTC1 | |
| Recombinant protein corresponding to Human KNTC1. Recombinant protein control fragment (Product #RP-91312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction