Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KNTC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588956 Supplier Invitrogen™ Supplier No. PA588956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, cytoskeleton, kinetochore, spindle Immunogen sequence: IDLIDREQGE DCLLLLKSLP PAEAEKTAER VIIWARLALQ EEPDHSKEGK AWRMSVAKTS VDILKILCDI QKDNLQKKDE CEEMLKLFKE VASLQENFEV FLSFEDYSNS SLVADLREQH IKAHEVAQAK HKPGSTPEPI AAEVRSPSME SKLHRQALAL QMSKQELEAE LTLRALKDGN IKTALKKCSD LFKYHCNADT G.

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KNTC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.70 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KNTC1
Gene Accession No. P50748
Gene Alias D330012D13; FLJ36151; HsROD; Kiaa0166; kinetochore associated 1; kinetochore-associated protein 1; Kinetochore-associated protein 1-like protein; KNTC1; LOW QUALITY PROTEIN: kinetochore-associated protein 1; ROD; Rough deal homolog; Rough Deal homolog, centromere/kinetochore protein
Gene Symbols KNTC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 900-1100 of human KNTC1 (NP_0555231).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304477, 9735
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.