Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KNTC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988028
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8028 Supplier Bioss Supplier No. BS7808R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.

Specifications

Antigen KNTC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KNTC1
Gene Accession No. P50748
Gene Alias BOS_16668; D330012D13; FLJ36151; HsROD; Kiaa0166; kinetochore associated 1; Kinetochore-associated protein 1; Kntc1; LOW QUALITY PROTEIN: kinetochore-associated protein 1; ROD; Rough deal homolog; Rough Deal homolog, centromere/kinetochore protein
Gene Symbols KNTC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KNTC1/Kinetochore.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9735
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.