Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungstic acid, 99+%

Catalog No. AC207615000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 7783-03-1 | H2O4W | 249.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-03-1
Molecular Formula H2O4W
Molecular Weight (g/mol) 249.86
MDL Number MFCD00011336
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
IUPAC Name dioxotungstendiol
SMILES O[W](O)(=O)=O

Specifications

Color Green-Yellow to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula H2WO4
Merck Index 15, 9998
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hf and alkalies
Formula Weight 249.86
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 5.5
Density 5.5000g/mL
Chemical Name or Material Tungstic acid
Show More Show Less
EINECSNumber 231-975-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.