Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Amber 203 Serum Vials
Shop All DWK Life Sciences Products

Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
30 mL
Height (Metric):
40 mm
47 mm
54 mm
86 mm
Diameter (Metric) Outer:
23 mm
30 mm
Description
- Serum vial with an aluminum seal finish
- Comes without a closure and is lighter in weight than molded bottles
- Manufactured from amber 203 borosilicate glass
- Enhanced by the most popular GPI aluminum seal finishes, this line offers a variety of choices
Specifications
Specifications
| Color | Amber |
| Material | Borosilicate Glass |
| Volume (Metric) | 10 mL |
| For Use With (Application) | Ideally suited for the handling, and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography samples |
| Material (Seal) | Aluminum |
| Volume (English) | 2.75 dr. |
| Autoclavable | Autoclavable |
| Height (Metric) | 54 mm |
| Certifications/Compliance | ASTM E438, Type I |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction