Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Packing Reservoir, with Joints
Shop All DWK Life Sciences Products
Description
- Reservoirs are available in a variety of sizes and joint configurations
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Type | Packing Reservoir with Joints |
| Volume (Metric) | 500 mL |