Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Graham Condensers

Outer joint at top and inner drip joint at the bottom.
Supplier: DWK Life Sciences 4390002425
Description
- Full length joints
- 24/40 joints
- No. 2 hose connections.

Specifications
392 mm | |
Borosilicate Glass | |
9.84 in. | |
24/40 |
15.43 in. | |
USP Type I, ASTM E438, Type I, Class A | |
Graham Condenser | |
250 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction