Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Single and Double Sealing Manifold
SDP
Catalog No. K8970470001 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Single NMR Sealing Manifold
For Use With (Equipment):
5 mm NMR Tube
Complete NMR Tube Sealing Manifold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
K897047-0001 Single NMR Sealing Manifold 5 mm NMR Tube
K897048-0001 Single NMR Sealing Manifold Complete NMR Tube Sealing Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K897047-0001 Supplier DWK Life Sciences Supplier No. 8970470001
Add to Cart
Add to Cart
Specifications
Product Type Single NMR Sealing Manifold
Includes Bevel Seal™ threaded No. 2 hose connection and cap with aluminum shield, Kontes™ Hi-Vac™ valves, viton O-ring, single NMR manifold
Material Borosilicate Glass
For Use With (Application) Provides an easy way to evacuate and tip-off NMR tubes without additional sealing to other glass apparatus
For Use With (Equipment) 5 mm NMR Tube