Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
SDP
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
Supplier:  DWK Life Sciences 7742400019
 View more versions of this product
Catalog No. K7742400019
Only null left
Add to Cart
Description
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
  • Leak-proof, air-tight seals
  • Wide top flange for ease of use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plug Stopper
Polyethylene
19/22 ,19/38
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.