Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
SDP
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
$40.80 - $72.60
Specifications
Material Polyethylene
Product Type Plug Stopper
View More Specs
Products 4
Catalog Number Mfr. No. Standard Taper Joint Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Price Quantity & Availability  
K7742400014
SDP
View Documents
DWK Life Sciences
7742400014
14/10, 14/20, 14/35
Case of 100 for $42.85
Only null left
Add to Cart
K7742400019
SDP
View Documents
DWK Life Sciences
7742400019
19/22 ,19/38
Case for $72.60
Only null left
Add to Cart
K7742400024
SDP
View Documents
DWK Life Sciences
7742400024
24/35, 24/40
Case of 100 for $40.80
Only null left
Add to Cart
K7742400029
SDP
View Documents
DWK Life Sciences
7742400029
29/26, 29/42
Case for $69.10
Only null left
Add to Cart
Description
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
  • Leak-proof, air-tight seals
  • Wide top flange for ease of use
Specifications
Specifications
Polyethylene
Plug Stopper
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.