Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Serum Vials KG-33 Borosilicate Glass with Aluminum Seal
SDP

Catalog No. p-313437 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (English):
0.5 dr.
2.75 dr.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
10-310-242A 2.75 dr.
03-422-19 0.5 dr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-310-242A Supplier DWK Life Sciences Supplier No. 62121D10
Only null left
Add to Cart
Add to Cart

Autoclavable Kimble™ serum vials are ideally suited for the handling, containment, and storage of a variety of liquids.

  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Autoclavable
    Aluminum seals, stoppers and septa are available separately
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement
TRUSTED_SUSTAINABILITY

Specifications

Classification A
Color Clear
Diameter (Metric) Outer 25 mm
Material Borosilicate Glass
Volume (Metric) 10 mL
For Use With (Application) Freeze drying applications
Material (Seal) Aluminum
Volume (English) 2.75 dr.
Certifications/Compliance ASTM E438, Type I, Class A
Height (Metric) 54 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.