Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Shop All DWK Life Sciences Products
Click to view available options
Size:
10/18
19
19/22
24/25
27
38
Description
- Sizes Standard Taper 10/18 through Standard Taper 19/22 are solid
- Sizes Standard Taper 24/25 and larger are hollow
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Material | Glass |
| Length (Metric) | 2.2 cm |
| For Use With (Equipment) | Flasks, mixing cylinders and separatory funnels |
| Size | 19/22 |
| Standard Taper Joint | 19/22 |
| Product Type | Solid Pennyhead Stopper |