Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
SDP
Catalog No. p-4909181
Click to view available options
Volume (Metric):
1000 mL (Funnel), 4000 mL (Flask)
300 mL (Funnel), 1000 mL (Flask)
500 mL (Funnel), 2000 mL (Flask)
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
  • The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
  • The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
  • Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
  • Joints are 40/35
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Microfiltration Assembly
Volume (Metric) 300 mL (Funnel), 1000 mL (Flask)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.