Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (90mm)
GSA_VASDP
Catalog No. p-4910434 Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
1 L
2 L
Designed for large volume filtration analysis of corrosive liquids or samples with high particulate loads that would tend to clog a 47 mm diameter filter
This unit is also recommended for filtering large volumes of HPLC solvents.
  • The ground glass connection eliminates the possibility of phthalate contamination that can occur when using silicone or neoprene stoppers
      Each apparatus is supplied with 1000mL funnel; 90mm, 40-60 micron porosity, fritted glass support base; anodized aluminum clamp; and filtration flask
    • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Microfiltration Assembly
Volume (Metric) 1 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.