Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOR Antibody (387301), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3895APCCY7
Description
KOR Monoclonal antibody specifically detects KOR in Human samples. It is validated for Flow CytometrySpecifications
KOR | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
387301 | |
Flow Cytometry | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
NS0 mouse myeloma cell line transfected with human KOR, Met1-Val380, Accession # P41145 | |
0.1 mL | |
GPCR, Phospho Specific, Signal Transduction | |
4986 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction