Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOR Antibody (387301), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3895L
Description
KOR Monoclonal antibody specifically detects KOR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
KOR | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
387301 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
NS0 mouse myeloma cell line transfected with human KOR, Met1-Val380, Accession # P41145 | |
0.1 mL | |
GPCR, Signal Transduction | |
4986 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction