Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOR Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $450.00
Specifications
Antigen | KOR |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018031
![]() |
Novus Biologicals
NB11079876SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB11079876
![]() |
Novus Biologicals
NB11079876 |
0.1 mg |
Each for $450.00
|
|
|||||
Description
KOR Polyclonal specifically detects KOR in Human, Rat samples. It is validated for Western Blot.Specifications
KOR | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
GPCR, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
4986 | |
Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
50 kDa |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
OPRK1 | |
IgG | |
Affinity Purified | |
Kappa Opioid Receptor |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title