Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KOR Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11079876PCP
Description
KOR Polyclonal antibody specifically detects KOR in Human, Rat samples. It is validated for Western BlotSpecifications
KOR | |
Polyclonal | |
Western Blot | |
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK | |
Synthetic peptide comprising residues 366-380 of the human KOR-1 protein. Sequence identity is 66% in mouse and rat. | |
0.1 mL | |
GPCR, Phospho Specific, Signal Transduction | |
4986 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction