Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kos1 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277929
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7929 Supplier Leinco Technologies Inc Supplier No. 28108100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kos1 Polyclonal antibody specifically detects Kos1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen Kos1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TNK1
Gene Accession No. Q99ML2
Gene Alias CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 421-434 (QNQVQSLPAGSAEP) of mouse Kos1 conjugated to KLH at the N-terminus.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83813
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.