Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kos1/Tnk1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300237PE
Description
Kos1/Tnk1 Polyclonal antibody specifically detects Kos1/Tnk1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Kos1/Tnk1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
CD38 negative kinase 1;Kinase of embryonic stem cells;MGC46193;non-receptor tyrosine kinase;Non-receptor tyrosine-protein kinase TNK1;TNK1;tyrosine kinase, non-receptor 1;tyrosine kinase, non-receptor, 1 | |
C-terminal residues 421-434 [QNQVQSLPAGSAEP] of the 47 kDa mouse Kos1/ Tnk1 protein. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction