Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KPL2/Sperm Flagellar 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981407
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1407 Supplier Bioss Supplier No. BS11488R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen KPL2/Sperm Flagellar 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPEF2
Gene Accession No. Q9C093
Gene Alias cancer/testis antigen 122; CT122; KIAA1770; KPL2; Protein KPL2; SPEF2; sperm flagellar 2; Sperm flagellar protein 2; testis tissue sperm-binding protein Li 47a
Gene Symbols SPEF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KPL2/Sperm Flagellar 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79925
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.