Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPNA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584596 Supplier Invitrogen™ Supplier No. PA584596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFKRRNVATA EEETEEEVMS DGGFHEAQIS NM Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

Recombination activating proteins RAG1 and RAG2 regulate and mediate VJ recombination, the process by which genes for immunoglobulins and T-cell receptors are generated. Several other ubiquitously expressed proteins are thought to be recruited in the recombination process. Among these are the genes affected in severe combined immune deficiency and genes involved in ds-DNA break repair. The protein encoded by this gene interacts with RAG1 and may play a role in VJ recombination. Two transcript variants, one protein-coding and the other not, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KPNA1
Gene Accession No. P52294
Gene Alias AW494490; IMA2; Importin alpha 2; importin alpha 5; Importin alpha P1; importin alpha-5; importin alpha-S1; importin subunit alpha-1; importin subunit alpha-5; Importin subunit alpha-5, N-terminally processed; importin-alpha-S1; IPOA 1; IPOA1; IPOA5; karyopherin (importin) alpha 1; Karyopherin A2; karyopherin alpha 1; karyopherin alpha 1 (importin alpha 5); karyopherin subunit alpha 1; karyopherin subunit alpha-1; karyopherin/importin alpha-1; KPNA1; KPNA2; m-importin-alpha-S1; mSRP1; NPI1; NPI-1; Nucleoprotein interactor 1; RAG cohort 1; RAG cohort protein 2; RCH 1; RCH1; RCH2; recombination activating gene cohort 2; SRP 1; SRP1; SRP1 alpha; SRP1-beta
Gene Symbols KPNA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KPNA1. Recombinant protein control fragment (Product #RP-105260).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.