Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KPNA3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001373
Description
KPNA3 Polyclonal specifically detects KPNA3 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
KPNA3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
hSRP1, importin alpha 4, Importin alpha Q2, importin alpha-3, importin subunit alpha-3, importin-alpha-Q2, IPOA4, karyopherin alpha 3 (importin alpha 4), Karyopherin subunit alpha-3, QIP2, SRP1, SRP1gamma, SRP1-gamma, SRP4 | |
Goat | |
Affinity Purified | |
RUO | |
3839 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.1-0.3 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 6 μg/mL, Peptide ELISA detection limit 1:128000 | |
NP_002258. | |
KPNA3 | |
Peptide with sequence C-DPTANLQTKEFNF corresponding to C-Terminus according to NP_002258.2. | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction