Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPNA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117127 Supplier Invitrogen™ Supplier No. PA5117127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: BT-474, Jurkat, A-549, U-251MG, HeLa Immunogen sequence: MAENPSLENH RIKSFKNKGR DVETMRRHRN EVTVELRKNK RDEHLLKKRN VPQEESLEDS DVDADFKAQN VTLEAILQNA TSDNPVVQLS AVQAARKLLS SDRNPPIDDL IKSGILPILV KCLERDDNPS LQFEAAWALT NIASGTSAQT QAVVQSNAVP LFLRLLRSPH QNVCEQAVWA LGNIIGDGPQ CRDYVISLGV VKPLLSFISP.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KPNA3
Gene Accession No. O00505, O35344
Gene Alias hSRP1; importin; importin alpha 4; importin alpha Q2; importin alpha-3; importin subunit alpha-3; importin subunit alpha-4; importin-alpha-Q2; IPOA4; karyopherin (importin) alpha 3; karyopherin alpha 3; karyopherin alpha 3 (importin alpha 4); karyopherin subunit alpha 3; karyopherin subunit alpha-3; Kpna3; predicted role in nuclear import; qip2; RP11-432M24.3; SRP1; SRP1gamma; SRP1-gamma; SRP4
Gene Symbols KPNA3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human KPNA3 (NP_0022582).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16648, 361055, 3839
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.