Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KPNA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145438
Description
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications
KPNA3 | |
Polyclonal | |
Unconjugated | |
KPNA3 | |
hSRP1; importin; importin alpha 4; importin alpha Q2; importin alpha-3; importin subunit alpha-3; importin subunit alpha-4; importin-alpha-Q2; IPOA4; karyopherin (importin) alpha 3; karyopherin alpha 3; karyopherin alpha 3 (importin alpha 4); karyopherin subunit alpha 3; karyopherin subunit alpha-3; Kpna3; predicted role in nuclear import; qip2; RP11-432M24.3; SRP1; SRP1gamma; SRP1-gamma; SRP4 | |
Rabbit | |
Protein A | |
RUO | |
16648, 361055, 3839, 396715 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
O00505, O35344 | |
KPNA3 | |
Recombinant Human KPNa3 protein, Glu3-Val220 (Accession #O00505). | |
20 μL | |
Primary | |
Human, Mouse, Rat, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction