Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KPNA4 Polyclonal Antibody
KPNA4 Polyclonal Antibody
Supplier: Thermo Scientific PA521033
Description
KPNA4 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
KPNA4 | |
Polyclonal | |
Unconjugated | |
KPNA4 | |
1110058D08Rik; importin; Importin alpha Q1; importin subunit alpha-3; importin subunit alpha-4; importin-alpha-Q1; IPOA3; karyopherin (importin) alpha 4; karyopherin alpha 4; karyopherin alpha 4 (importin alpha 3); karyopherin subunit alpha 4; karyopherin subunit alpha-4; KPNA4; MGC12217; MGC26703; predicted role in nuclear import; qip1; Qip1 protein; SRP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16649, 361959, 3840 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O00629, O35343 | |
KPNA4 | |
A 15 amino acid peptide near the amino terminus of human KPNA4. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction