Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544199
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544199 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544199 Supplier Invitrogen™ Supplier No. PA544199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TVMDSKIVQV ALNGLENILR LGEQESKQNG IGINPYCALI EEAYGLDKIE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 86%; Zebrafish: 93%.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC) which consists of 60-100 proteins and is probably 120 million daltons in molecular size. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion; larger molecules are transported by an active process. Most nuclear proteins contain short basic amino acid sequences known as nuclear localization signals (NLSs). KPNA5 protein belongs to the importin alpha protein family and is thought to be involved in NLS-dependent protein import into the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kpna5
Gene Accession No. O15131
Gene Alias hypothetical protein LOC553205; im:7151662; importin alpha 6; importin alpha-S2; Importin subunit alpha-6; Importin subunit alpha-6-like protein; importin subunit alpha-7; Ipoa6; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 5; karyopherin alpha 5 (importin alpha 6); karyopherin subunit alpha 5; karyopherin subunit alpha-5; Karyopherin subunit alpha-6; kpna5; Kpna6; NPI-2; SRP6; zgc:110662
Gene Symbols Kpna5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KPNA5 (aa 431-480).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3841
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.