Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KPNA5 Polyclonal Antibody
KPNA5 Polyclonal Antibody
Supplier: Thermo Scientific PA572750
Description
KPNA5 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Specifications
KPNA5 | |
Polyclonal | |
Unconjugated | |
Kpna5 | |
hypothetical protein LOC553205; im:7151662; importin alpha 6; importin alpha-S2; Importin subunit alpha-6; Importin subunit alpha-6-like protein; importin subunit alpha-7; Ipoa6; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 5; karyopherin alpha 5 (importin alpha 6); karyopherin subunit alpha 5; karyopherin subunit alpha-5; Karyopherin subunit alpha-6; kpna5; Kpna6; NPI-2; SRP6; zgc:110662 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O15131 | |
Kpna5 | |
KPNA5 antibody for an 18 amino acid synthetic peptide near the amino terminus of human KPNA5. The immunogen is within amino acids 50 - 100 of KPNA5. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction