Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPNA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115485 Supplier Invitrogen™ Supplier No. PA5115485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KPNA5.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC) which consists of 60-100 proteins and is probably 120 million daltons in molecular size. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion; larger molecules are transported by an active process. Most nuclear proteins contain short basic amino acid sequences known as nuclear localization signals (NLSs). KPNA5 protein belongs to the importin alpha protein family and is thought to be involved in NLS-dependent protein import into the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Kpna5
Gene Accession No. O15131, Q56R16
Gene Alias hypothetical protein LOC553205; im:7151662; importin alpha 6; importin alpha-S2; Importin subunit alpha-6; Importin subunit alpha-6-like protein; importin subunit alpha-7; Ipoa6; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 5; karyopherin alpha 5 (importin alpha 6); karyopherin subunit alpha 5; karyopherin subunit alpha-5; Karyopherin subunit alpha-6; kpna5; Kpna6; NPI-2; SRP6; zgc:110662
Gene Symbols Kpna5
Host Species Rabbit
Immunogen A synthesized peptide derived from human KPNA5(Accession O15131), corresponding to amino acid residues N222-V272.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294392, 3841
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.