Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPNA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597906 Supplier Invitrogen™ Supplier No. PA597906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KPNA6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Karyopherin, a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Ran-dependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA6 belongs to a subfamily within the KPNA family that also includes IPOA5&6. Down-regulation of KPNA6 by RNAi expression in HeLa cells strongly inhibited cell proliferation, possibly due to blocking the nuclear import of specific factors essential for cell growth and proliferation. KPNA6 also interacts with the Rev protein of HIV-1 and promote its nuclear import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPNA6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KPNA6
Gene Accession No. O60684
Gene Alias FLJ11249; importin alpha 7 subunit; importin alpha-S2; Importin subunit alpha-7; importin-alpha-S2; IPOA7; karyopherin (importin) alpha 5; karyopherin (importin) alpha 6; karyopherin alpha 6; karyopherin alpha 6 (importin alpha 7); karyopherin subunit alpha 6; karyopherin subunit alpha-6; Kpna5; Kpna6; KPNA7; MGC17918; NPI-2; RP4-622L5.1
Gene Symbols KPNA6
Host Species Rabbit
Immunogen Recombinant Human Importin subunit alpha-7 protein (277-536AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23633
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.