Learn More
Invitrogen™ KPRP Polyclonal Antibody

Description
Immunogen sequence: STQCQYQGSY SSCGPQFQSR ATCNNYTPQF QLRPSYSSCF PQYRSRTSFS PCVPQCQTQG SYGSFTEQHR SRSTSRCLPP PRRLQLFPRS CSPPRRFEP Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 52%.
Specifications
Specifications
| Antigen | KPRP |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KPRP |
| Gene Accession No. | Q5T749 |
| Gene Alias | C1orf45; hKPRP; keratinocyte expressed, proline-rich protein; keratinocyte proline rich protein; keratinocyte proline-rich protein; KPRP |
| Gene Symbols | KPRP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.