Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KPTN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559735 Supplier Invitrogen™ Supplier No. PA559735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLLSGNDPAI HLYKENEGLH QFEEQPVENL FPELTNLTSS VLWLDVHNFP GTSRRLSALG CQSGYVRVAH VDQRSREVLQ MWSVL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

Kaptin, also named as KPTN and Actin-associated protein 2E4, originally isolated from human blood platelets and likely to be involved in the actin rearrangements occurring during activation. It plays a unique role in the actin rearrangements that accompany platelet activation and stereocilia formation. Kaptin is necessary for normal neuromorphogenesis. It may play a role in producing the sensory apparatus in hair cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KPTN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KPTN
Gene Accession No. Q8VCX6, Q9Y664
Gene Alias 2310042D10Rik; 2E4; actin-associated protein 2E4; actin-binding protein; C030013F01Rik; kaptin; kaptin (actin binding protein); kaptin, actin binding protein; KICSTOR complex protein kaptin; KPTN; MRT41
Gene Symbols KPTN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KPTN. Recombinant protein control fragment (Product #RP-98988).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11133, 308107, 70394
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.