Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Krackeler Scientific Merlin Microseal™ Inlet Septa

Description
- Lifetime of more than 2000 injections, depending on samples and operating conditions
- Greatly reduced instrument downtime for septa changes and injection port liner changes due to septa particulates
- Two distinct sealing mechanisms: double O-ring type seal around the syringe needle and spring-assisted duckbill to seal the injection port
- Temperature range: 50° to 400°C
- Requires 23 gauge syringe needle
Specifications
Specifications
| For Use With (Application) | For split/splitless injection and SPME |
| Temperature Range (English) | 122°F to 752°F |
| Temperature Range (Metric) | 50°C to 400°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction