Learn More
KRAP Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304607A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SSFA2 gene ontology annotations related to this gene include actin binding.Specifications
KRAP | |
Polyclonal | |
Unconjugated | |
SSFA2 | |
CS-1, CS1, KRAP, SPAG13, KRAS-induced actin-interacting protein, cleavage signal-1 protein, ki-ras-induced actin-interacting protein, sperm associated antigen 13, sperm-specific antigen 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6744 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P28290 | |
SSFA2 | |
Between 1209 and 1259 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.